2-(Benzyloxy)-3-chloro-1-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52466-1G 52466-5G 52466-25G
77341-150EA 72.1 USD
77341-150 77365-042 77376-670
2-(Benzyloxy)-3-chloro-1-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-3-chloro-1-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530176
CAS Number: 2734779-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR