Methyl 2,4-difluoro-3-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

52463-500MG 52463-1G 52463-5G 52463-25G
77328-698EA 206 USD
77328-698 77352-378 77365-038 77387-130
Methyl 2,4-difluoro-3-isopropoxybenzoate ≥97%
Methyl 2,4-difluoro-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530174
CAS Number: 2384557-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR