3-Methyl-4-(pyridin-4-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52443-500MG 52443-1G 52443-5G
77336-040EA 374.92 USD
77336-040 77352-372 77376-656
3-Methyl-4-(pyridin-4-yl)benzaldehyde ≥97%
3-Methyl-4-(pyridin-4-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107851
CAS Number: 1507036-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR