N-(3-Bromo-5-methylpyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

52436-500MG 52436-1G 52436-5G 52436-25G
77328-692EA 187.46 USD
77328-692 77352-368 77365-028 77387-126
N-(3-Bromo-5-methylpyridin-2-yl)pivalamide ≥97%
N-(3-Bromo-5-methylpyridin-2-yl)pivalamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O
MW: 271.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404067
CAS Number: 850014-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR