N-(3-Chloro-5-methylpyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

52435-500MG 52435-1G 52435-5G 52435-25G
77328-690EA 225.57 USD
77328-690 77352-366 77365-026 77387-124
N-(3-Chloro-5-methylpyridin-2-yl)pivalamide ≥97%
N-(3-Chloro-5-methylpyridin-2-yl)pivalamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O
MW: 226.703 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404066
CAS Number: 2635937-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR