4'-Chloro-3'-methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52432-500MG 52432-1G 52432-5G 52432-25G
77328-688EA 111.24 USD
77328-688 77343-306 77365-024 77376-650
4'-Chloro-3'-methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Chloro-3'-methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₂
MW: 260.716 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25587792
CAS Number: 1513775-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR