2',3'-Dimethoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52431-500MG 52431-1G 52431-5G 52431-25G
77328-686EA 192.61 USD
77328-686 77352-362 77365-022 77387-122
2',3'-Dimethoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2',3'-Dimethoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25270243
CAS Number: 1551932-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR