3'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52430-500MG 52430-1G 52430-5G 52430-25G
77328-684EA 129.78 USD
77328-684 77344-312 77365-020 77376-648
3'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Methoxy-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25119582
CAS Number: 1506675-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR