3',5'-Difluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
52429-500MG
52429-1G
52429-5G
52429-25G
77328-682EA
146.26
USD
77328-682
77345-166
77365-018
77387-120
3',5'-Difluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',5'-Difluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₀F₂O MW: 232.225 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23570428 CAS Number: 1533942-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...