2'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

52427-500MG 52427-1G 52427-5G
77336-476EA 388.31 USD
77336-476 77352-358 77376-644
2'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%
2'-Chloro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD24107850
CAS Number: 1501484-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR