2'-Fluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52426-500MG 52426-1G 52426-5G
77336-474EA 388.31 USD
77336-474 77352-356 77376-642
2'-Fluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Fluoro-2-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO
MW: 214.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107868
CAS Number: 1516102-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR