(4-Fluoro-[1,1'-biphenyl]-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

52423-500MG 52423-1G 52423-5G
77328-678EA 318.27 USD
77328-678 77352-354 77376-640
(4-Fluoro-[1,1'-biphenyl]-2-yl)boronic acid ≥97%
(4-Fluoro-[1,1'-biphenyl]-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₂
MW: 216.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404064
CAS Number: 2243975-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR