5-Chloro-2-(o-tolyl)isonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

52420-500MG 52420-1G 52420-5G
77337-912EA 451.14 USD
77337-912 77352-352 77376-638
5-Chloro-2-(o-tolyl)isonicotinaldehyde ≥97%
5-Chloro-2-(o-tolyl)isonicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404063
CAS Number: 2635937-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR