5-Chloro-4-methyl-2-(o-tolyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

52419-500MG 52419-1G 52419-5G
77328-676EA 318.27 USD
77328-676 77352-350 77376-636
5-Chloro-4-methyl-2-(o-tolyl)pyridine ≥97%
5-Chloro-4-methyl-2-(o-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN
MW: 217.694 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404062
CAS Number: 2635937-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR