2,3-Dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

52416-500MG 52416-1G 52416-5G
77336-082EA 375.95 USD
77336-082 77352-346 77376-632
2,3-Dimethoxy-1,1'-biphenyl ≥97%
2,3-Dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404059
CAS Number: 82895-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR