3-Chloro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

52415-500MG 52415-1G 52415-5G
77337-510EA 416.12 USD
77337-510 77352-344 77376-630
3-Chloro-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
3-Chloro-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₈ClF₃
Storage Temperature: Ambient
MDL Number: MFCD14701715
CAS Number: 1214331-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR