2-(Benzyloxy)-5-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

52413-500MG 52413-1G 52413-5G
77328-674EA 324.45 USD
77328-674 77352-340 77376-626
2-(Benzyloxy)-5-fluoro-1,1'-biphenyl ≥97%
2-(Benzyloxy)-5-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₉H₁₅FO
MW: 278.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404058
CAS Number: 2624417-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR