3,5-Dichlorobiphenyl (PCB No. 14) ≥97%

Supplier: AOBChem USA

Warning

52411-500MG 52411-1G 52411-5G 52411-25G
77328-672EA 135.96 USD
77328-672 77344-512 77365-016 77387-118
3,5-Dichlorobiphenyl (PCB No. 14) ≥97%
3,5-Dichlorobiphenyl (PCB No. 14)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00055256
CAS Number: 34883-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR