3-Ethoxy-4-fluoro-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52398-500MG 52398-1G 52398-5G 52398-25G
77328-666EA 263.68 USD
77328-666 77352-332 77365-012 77387-112
3-Ethoxy-4-fluoro-5-(methylthio)benzoic acid ≥97%
3-Ethoxy-4-fluoro-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃S
Storage Temperature: Ambient
MDL Number: MFCD33404055
CAS Number: 2624417-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR