1,5-Dibromo-3-(tert-butyl)-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

52390-500MG 52390-1G 52390-5G 52390-25G
77328-662EA 182.31 USD
77328-662 77346-886 77365-008 77387-108
1,5-Dibromo-3-(tert-butyl)-2-methoxybenzene ≥97%
1,5-Dibromo-3-(tert-butyl)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄Br₂O
MW: 322.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22574056
CAS Number: 31918-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR