2-(Benzyloxy)-1,5-dibromo-3-(tert-butyl)benzene ≥97%

Supplier: AOBChem USA

Warning

52389-500MG 52389-1G 52389-5G 52389-25G
77328-660EA 281.19 USD
77328-660 77352-328 77365-006 77387-106
2-(Benzyloxy)-1,5-dibromo-3-(tert-butyl)benzene ≥97%
2-(Benzyloxy)-1,5-dibromo-3-(tert-butyl)benzene

Precaución: for Research Use Only

Formula: C₁₇H₁₈Br₂O
MW: 398.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564218
CAS Number: 2734775-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR