3-(tert-Butyl)-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52383-500MG 52383-1G 52383-5G 52383-25G
77328-658EA 223.51 USD
77328-658 77352-326 77365-004 77387-104
3-(tert-Butyl)-4-methoxybenzaldehyde ≥97%
3-(tert-Butyl)-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01104342
CAS Number: 107430-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR