3,3-Difluoro-1,5-diphenylpyrrolidin-2-one ≥97%

Supplier: AOBChem USA

Warning

52378-500MG 52378-1G 52378-5G
77338-646EA 506.76 USD
77338-646 77352-322 77376-618
3,3-Difluoro-1,5-diphenylpyrrolidin-2-one ≥97%
3,3-Difluoro-1,5-diphenylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₂NO
MW: 273.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404048
CAS Number: 1949728-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR