2-(tert-Butyl)-6-chlorophenol ≥97%

Supplier: AOBChem USA

Warning

52351-500MG 52351-1G 52351-5G 52351-25G
77328-656EA 189.52 USD
77328-656 77352-320 77365-002 77387-102
2-(tert-Butyl)-6-chlorophenol ≥97%
2-(tert-Butyl)-6-chlorophenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD16996889
CAS Number: 4237-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR