N-(5-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

52338-1G 52338-5G 52338-25G
77340-720EA 60.77 USD
77340-720 77364-994 77376-612
N-(5-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide ≥97%
N-(5-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO₂
MW: 298.057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14683657
CAS Number: 338446-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR