1-Fluoro-2,4-dimethylnaphthalene ≥97%

Supplier: AOBChem USA

Warning

52336-500MG 52336-1G 52336-5G
77339-300EA 573.71 USD
77339-300 77352-318 77376-610
1-Fluoro-2,4-dimethylnaphthalene ≥97%
1-Fluoro-2,4-dimethylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁F
MW: 174.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756906
CAS Number: 2901084-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR