Ethyl 2-bromo-4,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

52322-500MG 52322-1G 52322-5G
77338-540EA 497.49 USD
77338-540 77352-316 77376-608
Ethyl 2-bromo-4,5-dichlorobenzoate ≥97%
Ethyl 2-bromo-4,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrCl₂O₂
MW: 297.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32661566
CAS Number: 2169215-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR