Methyl 2-bromo-4,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

52321-500MG 52321-1G 52321-5G
77338-538EA 497.49 USD
77338-538 77352-314 77376-606
Methyl 2-bromo-4,5-dichlorobenzoate ≥97%
Methyl 2-bromo-4,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O₂
MW: 283.934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32661565
CAS Number: 2091686-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR