5'-Acetyl-2'-chloro-3-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52315-500MG 52315-1G 52315-5G
77336-924EA 400.67 USD
77336-924 77352-312 77376-604
5'-Acetyl-2'-chloro-3-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
5'-Acetyl-2'-chloro-3-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD33404040
CAS Number: 2624416-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR