3'-Chloro-3,4'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52314-500MG 52314-1G 52314-5G 52314-25G
77328-644EA 160.68 USD
77328-644 77345-796 77364-986 77387-092
3'-Chloro-3,4'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Chloro-3,4'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD33404039
CAS Number: 1994157-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR