3-Chloro-4-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52276-500MG 52276-1G 52276-5G
77338-140EA 466.59 USD
77338-140 77352-302 77376-594
3-Chloro-4-methoxy-5-methylbenzaldehyde ≥97%
3-Chloro-4-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246302
CAS Number: 62316-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR