(2-Fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52272-1G 52272-5G 52272-25G
77340-718EA 60.77 USD
77340-718 77364-980 77376-590
(2-Fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-4-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₄S
Storage Temperature: Ambient
MDL Number: MFCD22489519
CAS Number: 1823055-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR