3,5-Bis(trifluoromethyl)thioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)(methyl)sulphane

52261-1G 52261-5G 52261-25G
77340-716EA 60.77 USD
77340-716 77364-978 77376-588
3,5-Bis(trifluoromethyl)thioanisole ≥97%
3,5-Bis(trifluoromethyl)thioanisole

Precaución: for Research Use Only

Formula: C₉H₆F₆S
MW: 260.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789174
CAS Number: 685892-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR