(4-Fluoro-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

52259-1G 52259-5G 52259-25G
77341-772EA 104.03 USD
77341-772 77364-976 77376-586
(4-Fluoro-2-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(4-Fluoro-2-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₄S
MW: 210.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527243
CAS Number: 1379323-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR