3-Fluoro-2-hydroxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52258-500MG 52258-1G 52258-5G
77328-640EA 347.11 USD
77328-640 77352-300 77376-584
3-Fluoro-2-hydroxy-4-methylbenzoic acid ≥97%
3-Fluoro-2-hydroxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30305717
CAS Number: 1702914-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR