(4-Bromo-2-chlorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

52243-500MG 52243-1G 52243-5G 52243-25G
77328-638EA 87.55 USD
77328-638 77342-306 77364-972 77376-580
(4-Bromo-2-chlorophenyl)(thiomorpholino)methanone ≥97%
(4-Bromo-2-chlorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrClNOS
MW: 320.633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404027
CAS Number: 2624417-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR