(E)-5-bromo-2,4-difluorobenzaldehyde oxime ≥95%

Supplier: AOBChem USA

Warning

52226-1G 52226-5G 52226-25G
77341-340EA 82.4 USD
77341-340 77364-968 77376-576
(E)-5-bromo-2,4-difluorobenzaldehyde oxime ≥95%
(E)-5-bromo-2,4-difluorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₄BrF₂NO
MW: 236.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902995
CAS Number: 2586233-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR