3',4'-Dichloro-2-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52187-500MG 52187-1G 52187-5G
77337-146EA 408.91 USD
77337-146 77352-298 77376-574
3',4'-Dichloro-2-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',4'-Dichloro-2-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇Cl₂FO
MW: 269.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25119585
CAS Number: 1361720-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR