2,4'-Dichloro-3'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52186-500MG 52186-1G 52186-5G
77338-704EA 510.88 USD
77338-704 77352-296 77376-572
2,4'-Dichloro-3'-fluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2,4'-Dichloro-3'-fluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇Cl₂FO
MW: 269.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404025
CAS Number: 2624416-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR