2,4'-Difluoro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52184-500MG 52184-1G 52184-5G
77338-012EA 458.35 USD
77338-012 77352-292 77376-568
2,4'-Difluoro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2,4'-Difluoro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404023
CAS Number: 2624417-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR