2,4'-Dichloro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

52182-500MG 52182-1G 52182-5G 52182-25G
77328-632EA 265.74 USD
77328-632 77352-288 77364-964 77387-086
2,4'-Dichloro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2,4'-Dichloro-5-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂O
MW: 265.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902993
CAS Number: 2586126-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR