4-Fluoro-3-methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52168-500MG 52168-1G 52168-5G
77335-844EA 367.71 USD
77335-844 77352-276 77376-558
4-Fluoro-3-methoxy-5-(methylthio)benzaldehyde ≥97%
4-Fluoro-3-methoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902988
CAS Number: 2090518-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR