3-Fluoro-2-hydroxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52152-500MG 52152-1G 52152-5G
77338-958EA 536.63 USD
77338-958 77352-274 77376-554
3-Fluoro-2-hydroxy-4-methylbenzaldehyde ≥97%
3-Fluoro-2-hydroxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658251
CAS Number: 1283718-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR