2-Fluoro-3-methyl-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

52146-500MG 52146-1G 52146-5G
77335-548EA 357.41 USD
77335-548 77352-272 77376-552
2-Fluoro-3-methyl-6-(methylthio)phenol ≥97%
2-Fluoro-3-methyl-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FOS
MW: 172.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178617
CAS Number: 2643368-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR