2-(Benzyloxy)-1,5-dibromo-3-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52143-500MG 52143-1G 52143-5G 52143-25G
77328-624EA 140.08 USD
77328-624 77344-748 77364-956 77387-080
2-(Benzyloxy)-1,5-dibromo-3-fluoro-4-methylbenzene ≥97%
2-(Benzyloxy)-1,5-dibromo-3-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₁Br₂FO
MW: 374.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564214
CAS Number: 2734777-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR