1,5-Dibromo-3-fluoro-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52142-500MG 52142-1G 52142-5G 52142-25G
77328-622EA 116.39 USD
77328-622 77343-420 77364-954 77376-546
1,5-Dibromo-3-fluoro-2-isopropoxy-4-methylbenzene ≥97%
1,5-Dibromo-3-fluoro-2-isopropoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br₂FO
MW: 326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564213
CAS Number: 2734775-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR