4-(Benzyloxy)-3-fluoro-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52117-500MG 52117-1G 52117-5G
77336-900EA 398.61 USD
77336-900 77352-268 77376-544
4-(Benzyloxy)-3-fluoro-2-(methylthio)benzoic acid ≥97%
4-(Benzyloxy)-3-fluoro-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃S
Storage Temperature: Ambient
MDL Number: MFCD33404022
CAS Number: 2624417-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR