2-Fluoro-3,5-dimethylphenol ≥95%

Supplier: AOBChem USA

Warning

52110-500MG 52110-1G 52110-5G
77337-246EA 409.94 USD
77337-246 77352-262 77376-538
2-Fluoro-3,5-dimethylphenol ≥95%
2-Fluoro-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23111065
CAS Number: 1253214-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR