2-Fluoro-3-hydroxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52108-500MG 52108-1G 52108-5G
77337-418EA 414.06 USD
77337-418 77352-260 77376-536
2-Fluoro-3-hydroxy-5-methylbenzaldehyde ≥97%
2-Fluoro-3-hydroxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD28502878
CAS Number: 1780209-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR