2,3-Dibromo-4-chloroaniline ≥97%

Supplier: AOBChem USA

Warning

52103-500MG 52103-1G 52103-5G 52103-25G
77328-620EA 161.71 USD
77328-620 77345-882 77364-952 77387-078
2,3-Dibromo-4-chloroaniline ≥97%
2,3-Dibromo-4-chloroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂ClN
Storage Temperature: Ambient
MDL Number: MFCD32902987
CAS Number: 96558-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR